Morpholin-3-ylmethanol ≥98%

Supplier: Ambeed
Warning

A908640-1G A908640-5G A908640-10G A908640-250MG
77603-270EA 70.73 USD
77603-270 77670-338 77873-361 77873-362
Morpholin-3-ylmethanol ≥98%
Morpholin-3-ylmethanol

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MDL Number: MFCD04038650
CAS Number: 106910-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR