Methyl 5-fluoro-1H-indazole-6-carboxylate ≥97%

Supplier: Ambeed
Warning

A908069-5G A908069-1G
77599-250EA 720.58 USD
77599-250 77618-540
Methyl 5-fluoro-1H-indazole-6-carboxylate ≥97%
Methyl 5-fluoro-1H-indazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇FN₂O₂
MW: 194.1625 g/mol
MDL Number: MFCD16875735
CAS Number: 1227270-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR