(S)-3-Amino-N-((1R,2S,3S,4R,5S)-5-amino-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-2-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxypropanamide sulf ≥98%

Supplier: Ambeed
Warning

A907713-5MG A907713-1MG A907713-50MG
77595-464EA 30.75 USD
77595-464 77670-336 77880-734
(S)-3-Amino-N-((1R,2S,3S,4R,5S)-5-amino-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-2-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxypropanamide sulf ≥98%
(S)-3-Amino-N-((1R,2S,3S,4R,5S)-5-amino-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-2-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxypropanamide sul

Precaución: for Research Use Only

Formula: C₂₂H₄₅N₅O₁₆S
MDL Number: MFCD01683537
CAS Number: 67814-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR