Methyl 5-chloro-2-formylbenzoate ≥95%

Supplier: Ambeed
Warning

A906546-1G A906546-250MG A906546-5G
77595-458EA 70.73 USD
77595-458 77618-538 77749-723
Methyl 5-chloro-2-formylbenzoate ≥95%
Methyl 5-chloro-2-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₇ClO₃
MDL Number: MFCD18382526
CAS Number: 1203589-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR