((2-(2-Bromoethoxy)ethoxy)methyl)benzene ≥96%

Supplier: Ambeed
Danger

A906529-1G A906529-5G
77599-248EA 76.88 USD
77599-248 77618-536
((2-(2-Bromoethoxy)ethoxy)methyl)benzene ≥96%
((2-(2-Bromoethoxy)ethoxy)methyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO₂
MW: 259.1400 g/mol
MDL Number: MFCD25977543
CAS Number: 125562-32-5
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR