5-(6-Hydroxybenzofuran-2-yl)benzene-1,3-diol ≥98%

Supplier: Ambeed
Warning

A904366-5MG A904366-100MG A904366-10MG A904366-50MG
77686-945EA 68 USD
77686-945 77893-698 77893-699 77893-700
5-(6-Hydroxybenzofuran-2-yl)benzene-1,3-diol ≥98%
5-(6-Hydroxybenzofuran-2-yl)benzene-1,3-diol

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₄
MDL Number: MFCD17166969
CAS Number: 56317-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR