4-Cyclopropylphenylboronic Acid Pinacol Ester ≥95%

Supplier: Ambeed
Warning

A903464-1G A903464-5G A903464-100MG A903464-250MG
77614-622EA 89.61 USD
77614-622 77886-904 77886-902 77886-903
4-Cyclopropylphenylboronic Acid Pinacol Ester ≥95%
4-Cyclopropylphenylboronic Acid Pinacol Ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₂
MDL Number: MFCD10698529
CAS Number: 1219741-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR