Iridium(IV) oxide 99.9% (metals basis) Ir ≥84.5%

Supplier: Ambeed
Danger

Synonyms: Iridium dioxide, Iridium(IV) oxide

A903419-100MG A903419-1G A903419-250MG
77603-256EA 51.25 USD
77603-256 77603-258 77603-260
Iridium(IV) oxide 99.9% (metals basis) Ir ≥84.5%
Iridium(IV) oxide

Precaución: for Research Use Only

Formula: IrO₂
MW: 224.22 g/mol
MDL Number: MFCD00011065
CAS Number: 12030-49-8
UN: 1479
ADR: 5.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR