5-Bromo-3-(difluoromethyl)-2-fluoropyridine ≥95%

Supplier: Ambeed
Warning

A903209-250MG A903209-100MG A903209-1G
77595-454EA 21.53 USD
77595-454 77749-714 77749-715
5-Bromo-3-(difluoromethyl)-2-fluoropyridine ≥95%
5-Bromo-3-(difluoromethyl)-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₆H₃BrF₃N
MDL Number: MFCD25477707
CAS Number: 1805222-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR