2-Bromo-5-iodothiophene ≥98%

Supplier: Ambeed
Warning

A903031-100MG A903031-5G
77684-860EA 12 USD
77684-860 77684-861
2-Bromo-5-iodothiophene ≥98%
2-Bromo-5-iodothiophene

Precaución: For research use only.

Formula: C₄H₂BrIS
MW: 288.932 g/mol
MDL Number: MFCD08695058
CAS Number: 29504-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR