4,4',4'',4'''-(21H,23H-Porphine-5,10,15,20-tetrayl)tetrakis[benzaldehyde] ≥95%

Supplier: Ambeed
Warning

A902818-50MG A902818-250MG
77623-132EA 202.91 USD
77623-132 77784-341
4,4',4'',4'''-(21H,23H-Porphine-5,10,15,20-tetrayl)tetrakis[benzaldehyde] ≥95%
4,4',4'',4'''-(21H,23H-Porphine-5,10,15,20-tetrayl)tetrakis[benzaldehyde]

Precaución: for Research Use Only

Formula: C₄₈H₃₀N₄O₄
CAS Number: 150805-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR