(E)-(4-Cyanostyryl)boronic acid ≥97%

Supplier: Ambeed
Warning

A902690-1G A902690-5G A902690-250MG
77784-338EA 391 USD
77784-338 77784-340 77784-339
(E)-(4-Cyanostyryl)boronic acid ≥97%

Precaución: for Research Use Only

Formula: C₉H₈BNO₂
CAS Number: 352525-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR