(R)-1-(2, 4-Difluorophenyl)ethylamine ≥96%

Supplier: Ambeed
Danger

Synonyms: (R)-1-(2,4-Difluorophenyl)ethanamine, (1R)-1-(2,4-difluorophenyl)ethanamine

A902575-250MG A902575-1G
77747-668EA 45 USD
77747-668 77747-667
(R)-1-(2, 4-Difluorophenyl)ethylamine ≥96%
(R)-1-(2, 4-Difluorophenyl)ethylamine

Precaución: for Research Use Only

Formula: C₈H₉F₂N
MW: 157.161 g/mol
MDL Number: MFCD06761833
CAS Number: 791098-84-5
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR