Polyethylene glycol 10,000

Supplier: Ambeed
Warning

A902461-500G A902461-100G
77573-816EA 6 USD
77573-816 77618-530
Polyethylene glycol 10,000
Polyethylene glycol 10,000

Precaución: for Research Use Only

Formula: H(OCH₂CH₂)nOH
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR