Boc-D-Orn(fmoc)-OH ≥95%

Supplier: Ambeed
Warning

Synonyms: (R)-5-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)pentanoic acid

A901705-5G A901705-250MG
77686-943EA 617 USD
77686-943 77901-867
Boc-D-Orn(fmoc)-OH ≥95%
Boc-D-Orn(fmoc)-OH

Precaución: for Research Use Only

Formula: C₂₅H₃₀N₂O₆
MDL Number: MFCD00151883
CAS Number: 163336-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR