(R)-2,5,7,8-Tetramethyl-2-((3E,7E)-4,8,12-trimethyltrideca-3,7,11-trien-1-yl)chroman-6-ol ≥95%

Supplier: Ambeed
Warning

Synonyms: α-Tocotrienol

A901293-100MG A901293-50MG
77893-694EA 758 USD
77893-694 77893-695
(R)-2,5,7,8-Tetramethyl-2-((3E,7E)-4,8,12-trimethyltrideca-3,7,11-trien-1-yl)chroman-6-ol ≥95%
(R)-2,5,7,8-Tetramethyl-2-((3E,7E)-4,8,12-trimethyltrideca-3,7,11-trien-1-yl)chroman-6-ol

Precaución: for Research Use Only

Formula: C₂₉H₄₄O₂
MDL Number: MFCD11045308
CAS Number: 58864-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR