1-((9H-Fluoren-2-yl)methyl)-1-cycloheptyl-3-mesitylurea ≥98%

Supplier: Ambeed
Warning

Synonyms: YM-750

A901198-1MG A901198-5MG A901198-10MG
77599-244EA 38.95 USD
77599-244 77599-246 77599-242
1-((9H-Fluoren-2-yl)methyl)-1-cycloheptyl-3-mesitylurea ≥98%
1-((9H-Fluoren-2-yl)methyl)-1-cycloheptyl-3-mesitylurea

Precaución: for Research Use Only

Formula: C₃₁H₃₆N₂O
MDL Number: MFCD00905424
CAS Number: 138046-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR