Fmoc-L-selenomethionine ≥95%

Supplier: Ambeed
Danger

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(methylselanyl)butanoic acid, Fmoc-SeMet-OH

A900610-100MG A900610-1G A900610-250MG
77893-691EA 53 USD
77893-691 77893-692 77893-693
Fmoc-L-selenomethionine ≥95%
Fmoc-L-selenomethionine

Precaución: for Research Use Only

Formula: C₂₀H₂₁NO₄Se
MDL Number: MFCD03093662
CAS Number: 1217852-49-7
UN: 3283
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR