(1R,5S)-3-Methyl-3,4,5,6-tetrahydro-1H-1,5-methanopyrido[1,2-a][1,5]diazocin-8(2H)-one ≥98%

Supplier: Ambeed
Warning

A900570-100MG A900570-50MG
77880-717EA 200 USD
77880-717 77880-718
(1R,5S)-3-Methyl-3,4,5,6-tetrahydro-1H-1,5-methanopyrido[1,2-a][1,5]diazocin-8(2H)-one ≥98%
(1R,5S)-3-Methyl-3,4,5,6-tetrahydro-1H-1,5-methanopyrido[1,2-a][1,5]diazocin-8(2H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O
MDL Number: MFCD00211091
CAS Number: 486-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR