Sodium tetradecyl sulfate ≥98%

Supplier: Ambeed
Warning

Synonyms: Tetradecyl sulphuric acid salt, Tetradecyl sulphate sodium, Sodium 1-tetradecyl sulphate

A900555-1G A900555-250MG
77618-524EA 58.71 USD
77618-524 77618-526
Sodium tetradecyl sulfate ≥98%
Sodium tetradecyl sulphate

Precaución: for Research Use Only

Formula: C₁₄H₂₉NaO₄S
MW: 316.44 g/mol
MDL Number: MFCD00007468
CAS Number: 1191-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR