(2R,2'R,3R,3'R)-2,2'-(3,4,6-trihydroxy-5-oxo-5H-benzo[7]annulene-1,8-diyl)bis(5,7-dihydroxychroman-3,2-diyl) bis(3,4,5-trihydroxybenzoate) ≥98%

Supplier: Ambeed
Warning

Synonyms: Theaflavin 3,3'-digallate

A900282-1MG A900282-10MG A900282-5MG A900282-100MG A900282-50MG
77603-248EA 51.25 USD
77603-248 77670-316 77670-318 77873-350 77873-351
(2R,2'R,3R,3'R)-2,2'-(3,4,6-trihydroxy-5-oxo-5H-benzo[7]annulene-1,8-diyl)bis(5,7-dihydroxychroman-3,2-diyl) bis(3,4,5-trihydroxybenzoate) ≥98%
(2R,2'R,3R,3'R)-2,2'-(3,4,6-trihydroxy-5-oxo-5H-benzo[7]annulene-1,8-diyl)bis(5,7-dihydroxychroman-3,2-diyl) bis(3,4,5-trihydroxybenzoate)

Precaución: for Research Use Only

Formula: C₄₃H₃₂O₂₀
MDL Number: MFCD06797365
CAS Number: 30462-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR