3-(9-(tert-Butyl)-8-((1-methyl-1H-1,2,4-triazol-5-yl)methoxy)pyrazolo[1,5-d][1,2,4]triazin-2-yl)-5-methylisoxazole ≥98%

Supplier: Ambeed
Warning

A899895-1MG A899895-5MG A899895-10MG
77618-516EA 40.17 USD
77618-516 77747-665 77747-664
3-(9-(tert-Butyl)-8-((1-methyl-1H-1,2,4-triazol-5-yl)methoxy)pyrazolo[1,5-d][1,2,4]triazin-2-yl)-5-methylisoxazole ≥98%
3-(9-(tert-Butyl)-8-((1-methyl-1H-1,2,4-triazol-5-yl)methoxy)pyrazolo[1,5-d][1,2,4]triazin-2-yl)-5-methylisoxazole

Precaución: for Research Use Only

Formula: C₁₇H₂₀N₈O₂
MDL Number: MFCD18086893
CAS Number: 342652-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR