Benzyl (2-(2-(2-hydroxyethoxy)ethoxy)ethyl)carbamate ≥97%

Supplier: Ambeed
Warning

A898378-1G A898378-250MG A898378-5G
77747-659EA 240 USD
77747-659 77747-660 77747-661
Benzyl (2-(2-(2-hydroxyethoxy)ethoxy)ethyl)carbamate ≥97%
Benzyl (2-(2-(2-hydroxyethoxy)ethoxy)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₅
MW: 283.32 g/mol
MDL Number: MFCD07357495
CAS Number: 205535-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR