Phenyl-3,4,6-tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-β-D-glucopyranoside ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,3S,4R,5R,6S)-2-(Acetoxymethyl)-6-(phenylthio)-5-(((2,2,2-trichloroethoxy)carbonyl)amino)tetrahydro-2H-pyran-3,4-diyl diacetate, Phenyl 3,4,6-Tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-β-D-glucopyranoside
A898156-5G
A898156-250MG
77595-442EA
111.73
USD
77595-442
77749-705
Phenyl-3,4,6-tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-β-D-glucopyranoside ≥98%
Phenyl-3,4,6-tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-beta-D-glucopyranoside
Precaución: for Research Use Only
Formula:
C₂₁H₂₄Cl₃NO₉S |
MDL Number:
MFCD11112181 CAS Number: 187022-49-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...