1-Methyl-1H-indazole-7-boronic acid ≥98%

Supplier: Ambeed
Warning

A898150-250MG A898150-1G A898150-5G
77670-314EA 32.23 USD
77670-314 77670-312 77873-347
1-Methyl-1H-indazole-7-boronic acid ≥98%
1-Methyl-1H-indazole-7-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MDL Number: MFCD09870054
CAS Number: 1001907-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR