(1R,2S)-2-(7-Hydroxy-5-phenylpyrazolo[1,5-a]pyrimidine-3-carboxamido)cyclohexanecarboxylic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: PF-06928215

A897530-1MG A897530-5MG A897530-10MG A897530-50MG
77623-124EA 120.51 USD
77623-124 77623-126 77784-330 77784-331
(1R,2S)-2-(7-Hydroxy-5-phenylpyrazolo[1,5-a]pyrimidine-3-carboxamido)cyclohexanecarboxylic acid ≥98%
(1R,2S)-2-(7-Hydroxy-5-phenylpyrazolo[1,5-a]pyrimidine-3-carboxamido)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₄O₄
MDL Number: MFCD31697699
CAS Number: 2378173-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR