(E)-2-(7-Fluoro-1-isobutyl-2-oxoindolin-3-ylidene)acetic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: ASP 7663, (2E)-2-[7-Fluoro-1,2-dihydro-1-(2-methylpropyl)-2-oxo-3H-indol-3-ylidene]acetic acid, ASP7663, (E)-[7-Fluoro-1-(2-methylpropyl)-2-oxo-1,2-dihydro-3H-indol-3-ylidene]acetic acid, (2E)-2-[7-Fluoro-1,2-dihydro-1-(2-methylpropyl)-2-oxo-3H-indol-3-ylidene]acetic acid

A897050-10MG A897050-1MG A897050-5MG
77618-508EA 135.96 USD
77618-508 77618-510 77618-512
(E)-2-(7-Fluoro-1-isobutyl-2-oxoindolin-3-ylidene)acetic acid ≥98%
(E)-2-(7-Fluoro-1-isobutyl-2-oxoindolin-3-ylidene)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄FNO₃
MW: 263.26 g/mol
MDL Number: MFCD29079014
CAS Number: 1190217-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR