2,3-Dimethoxy-5-pyridineboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dimethoxypyridine-5-boronic acid, pinacol ester, 2,3-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A896711-100MG A896711-1G A896711-250MG A896711-5G
77873-343EA 69 USD
77873-343 77873-344 77873-345 77873-346
2,3-Dimethoxy-5-pyridineboronic acid pinacol ester ≥98%
2,3-Dimethoxy-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₄
MDL Number: MFCD11857651
CAS Number: 1083168-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR