2,3,4-Trimethoxybenzyl alcohol ≥95%

Supplier: Ambeed
Warning

Synonyms: TriMetazidine EP Impurity D, (2,3,4-Trimethoxyphenyl)methanol

A895374-5G A895374-25G A895374-1G
77747-655EA 33 USD
77747-655 77747-654 77747-653
2,3,4-Trimethoxybenzyl alcohol ≥95%
2,3,4-Trimethoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₄
MDL Number: MFCD00004613
CAS Number: 71989-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR