2,3-Dimethylnaphthalene ≥98%

Supplier: Ambeed
Warning

A893860-100MG A893860-1G A893860-250MG A893860-5G
77880-712EA 24 USD
77880-712 77880-713 77880-714 77880-715
2,3-Dimethylnaphthalene ≥98%
2,3-Dimethylnaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₂
MDL Number: MFCD00004119
CAS Number: 581-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR