2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)ethanamine hydrochloride ≥95%
Supplier: Ambeed
|
Warning
|
A892871-250MG
A892871-1G
77749-704EA
42
USD
77749-704
77749-703
2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)ethanamine hydrochloride ≥95%
2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)ethanamine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₉Cl₂F₃N₂ MW: 261.0717 g/mol |
MDL Number:
MFCD31714214 CAS Number: 326816-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...