4-Benzyloxy-2-butanone ≥96%

Supplier: Ambeed
Warning

A892711-25G A892711-5G A892711-1G
77749-701EA 209 USD
77749-701 77749-702 77749-700
4-Benzyloxy-2-butanone ≥96%
4-Benzyloxy-2-butanone

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.22 g/mol
MDL Number: MFCD00042852
CAS Number: 6278-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR