4-Bromo-1-(4-fluorophenyl)-1H-imidazole ≥95%

Supplier: Ambeed
Warning

A892477-100MG A892477-1G A892477-250MG
77886-891EA 26 USD
77886-891 77886-892 77886-893
4-Bromo-1-(4-fluorophenyl)-1H-imidazole ≥95%
4-Bromo-1-(4-fluorophenyl)-1H-imidazole

Precaución: for Research Use Only

Formula: C₉H₆BrFN₂
MW: 241.06 g/mol
MDL Number: MFCD14706293
CAS Number: 623577-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR