3-Methoxynaphthalen-2-ol ≥97%

Supplier: Ambeed
Danger

A892337-250MG A892337-1G
77784-325EA 52 USD
77784-325 77784-324
3-Methoxynaphthalen-2-ol ≥97%
3-Methoxynaphthalen-2-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
MDL Number: MFCD00029345
CAS Number: 18515-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR