(4aR,5aR)-4,4a,5,5a-Tetrahydro-1H-cyclopropa[4,5]cyclopenta[1,2-c]pyrazole-3-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A891114-10MG A891114-1MG A891114-5MG
77599-234EA 132.23 USD
77599-234 77599-236 77599-238
(4aR,5aR)-4,4a,5,5a-Tetrahydro-1H-cyclopropa[4,5]cyclopenta[1,2-c]pyrazole-3-carboxylic acid ≥98%
(4aR,5aR)-4,4a,5,5a-Tetrahydro-1H-cyclopropa[4,5]cyclopenta[1,2-c]pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₈N₂O₂
MDL Number: MFCD27991272
CAS Number: 1268882-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR