5-Bromo-4-methylnicotinic acid ≥98%
Supplier: Ambeed
|
Warning
|
A890698-250MG
A890698-5G
77684-851EA
19
USD
77684-851
77684-852
5-Bromo-4-methylnicotinic acid ≥98%
5-Bromo-4-methylnicotinic acid
Precaución: For research use only.
Formula:
C₇H₆BrNO₂ MW: 216.03 g/mol |
MDL Number:
MFCD07439973 CAS Number: 677702-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...