2-Bromo-3,6-difluorophenol ≥97%

Supplier: Ambeed
Warning

A890510-1G A890510-25G
77618-496EA 34.2 USD
77618-496 77618-498
2-Bromo-3,6-difluorophenol ≥97%
2-Bromo-3,6-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
MDL Number: MFCD13176518
CAS Number: 1208077-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR