3-Iodo-N,N-dimethylaniline ≥95%

Supplier: Ambeed
Warning

A890322-1G A890322-100MG A890322-5G
77749-698EA 254 USD
77749-698 77749-697 77749-699
3-Iodo-N,N-dimethylaniline ≥95%
3-Iodo-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀IN
MW: 247.08 g/mol
MDL Number: MFCD24133896
CAS Number: 33454-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR