(+/-)-2-(Trifluoromethyl)piperazine ≥98%

Supplier: Ambeed
Danger

Synonyms: 2-(Trifluoromethyl)piperazine

A889340-250MG A889340-100MG A889340-10G A889340-1G A889340-5G
77674-320EA 43 USD
77674-320 77882-804 77882-805 77882-806 77882-807
(+/-)-2-(Trifluoromethyl)piperazine ≥98%
(+/-)-2-(Trifluoromethyl)piperazine

Precaución: for Research Use Only

Formula: C₅H₉F₃N₂
MDL Number: MFCD07373382
CAS Number: 131922-05-9
UN: 3263
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR