(R)-4-Hydroxy-5-(octyloxy)-5-oxopentanoic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: (2R)-Octyl-α-hydroxyglutarate

A888839-10MG A888839-1MG A888839-5MG
77599-222EA 106.6 USD
77599-222 77599-224 77599-226
(R)-4-Hydroxy-5-(octyloxy)-5-oxopentanoic acid ≥97%
(R)-4-Hydroxy-5-(octyloxy)-5-oxopentanoic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₄O₅
MDL Number: MFCD30735809
CAS Number: 1391194-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR