RO8191 ≥90%

Supplier: Ambeed
Warning

Synonyms: RO4948191, 2-[2,4-Bis(trifluoromethyl)imidazo[1,2-a]1,8-naphthyridin-8-yl]-1,3,4-oxadiazole, 8-(1,3,4-Oxadiazol-2-yl)-2,4-bis(trifluoromethyl)-imidazo[1,2-a][1,8]naphthyridine

A888527-250MG A888527-100MG A888527-5G
77599-218EA 100.45 USD
77599-218 77623-106 77623-108
RO8191 ≥90%
RO8191

Precaución: for Research Use Only

Formula: C₁₄H₅F₆N₅O
MW: 373.21 g/mol
MDL Number: MFCD03102493
CAS Number: 691868-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR