3-(p-Tolyl)pyridine ≥97%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Methylphenyl)pyridine

A888374-250MG A888374-1G
77618-494EA 20.6 USD
77618-494 77749-694
3-(p-Tolyl)pyridine ≥97%
3-(p-Tolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁N
MW: 169.222 g/mol
MDL Number: MFCD04038775
CAS Number: 4423-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR