(E)-1-((E)-4-((E)-5-Carbamoyl-2-((1-ethyl-3-methyl-1H-pyrazole-5-carbonyl)imino)-7-(3-morpholinopropoxy)-2,3-dihydro-1H-benzo[d]imidazol-1-yl)but-2-en-1-yl)-2-((1-ethyl-3-methyl-1H-pyrazole-5-carbonyl)imino)-7-methoxy-2,3-dihydro-1H-benzo[d]imidazole-5-carboxamide ≥99%

Supplier: Ambeed
Warning

A885666-5MG A885666-1MG A885666-2MG A885666-10MG
77618-482EA 385.22 USD
77618-482 77618-480 77749-693 77749-692
(E)-1-((E)-4-((E)-5-Carbamoyl-2-((1-ethyl-3-methyl-1H-pyrazole-5-carbonyl)imino)-7-(3-morpholinopropoxy)-2,3-dihydro-1H-benzo[d]imidazol-1-yl)but-2-en-1-yl)-2-((1-ethyl-3-methyl-1H-pyrazole-5-carbonyl)imino)-7-methoxy-2,3-dihydro-1H-benzo[d]imidazole-5-carboxamide ≥99%
(E)-1-((E)-4-((E)-5-Carbamoyl-2-((1-ethyl-3-methyl-1H-pyrazole-5-carbonyl)imino)-7-(3-morpholinopropoxy)-2,3-dihydro-1H-benzo[d]imidazol-1-yl)but-2-en-1-yl)-2-((1-ethyl-3-methyl-1H-pyrazole-5-carbonyl)imino)-7-methoxy-2,3-dihydro-1H-benzo[d]imidazole-5-c

Precaución: for Research Use Only

Formula: C₄₂H₅₁N₁₃O₇
MW: 849.94 g/mol
CAS Number: 2138498-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR