1-Oxa-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester ≥95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 1-oxa-7-azaspiro[3.5]nonane-7-carboxylate

A885142-1G A885142-5G
77884-831EA 40 USD
77884-831 77884-832
1-Oxa-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester ≥95%
1-Oxa-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.3 g/mol
MDL Number: MFCD08706105
CAS Number: 864684-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR