Diethyl 3-Cyclopentene-1,1-dicarboxylate ≥98%

Supplier: Ambeed
Warning

A884829-5G A884829-1G A884829-25G
77573-788EA 66 USD
77573-788 77891-657 77891-658
Diethyl 3-Cyclopentene-1,1-dicarboxylate ≥98%
Diethyl 3-Cyclopentene-1,1-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₄
MDL Number: MFCD04115567
CAS Number: 21622-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR