Cobalt(II) acetylacetonate hydrate ≥98%

Supplier: Ambeed
Danger

A884172-100G A884172-25G A884172-500G
77603-214EA 19.48 USD
77603-214 77603-216 77603-218
Cobalt(II) acetylacetonate hydrate ≥98%
Cobalt(II) acetylacetonate hydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₄CoO₄·xH₂O
MDL Number: MFCD00149056
CAS Number: 123334-29-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR