Methyl isoquinoline-6-carboxylate ≥98%

Supplier: Ambeed
Warning

A883786-1G A883786-250MG A883786-5G
77674-314EA 100 USD
77674-314 77884-828 77884-829
Methyl isoquinoline-6-carboxylate ≥98%
Methyl isoquinoline-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MDL Number: MFCD11226961
CAS Number: 173089-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR