3-Chloro-2-(2-methylpropoxy)pyridine ≥95%

Supplier: Ambeed
Warning

A883657-100MG A883657-1G A883657-250MG
77901-621EA 47 USD
77901-621 77902-042 77900-898
3-Chloro-2-(2-methylpropoxy)pyridine ≥95%
3-Chloro-2-(2-methylpropoxy)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO
MDL Number: MFCD21609702
CAS Number: 1288989-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR