Monobenzyl malonate ≥97%

Supplier: Ambeed
Warning

A883205-10G A883205-5G A883205-1G
77595-432EA 60.48 USD
77595-432 77603-208 77627-704
Monobenzyl malonate ≥97%
Monobenzyl malonate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD00138132
CAS Number: 40204-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR