(E)-N1-(Adamantan-2-yl)-N2-(3,7-dimethylocta-2,6-dien-1-yl)ethane-1,2-diamine ≥98%

Supplier: Ambeed
Warning

A881317-10MG A881317-1MG A881317-5MG
77618-466EA 207.21 USD
77618-466 77618-468 77618-470
(E)-N1-(Adamantan-2-yl)-N2-(3,7-dimethylocta-2,6-dien-1-yl)ethane-1,2-diamine ≥98%
(E)-N1-(Adamantan-2-yl)-N2-(3,7-dimethylocta-2,6-dien-1-yl)ethane-1,2-diamine

Precaución: for Research Use Only

Formula: C₂₂H₃₈N₂
MDL Number: MFCD11046358
CAS Number: 502487-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR